Please login to your online account to display your discounted pricing

anti-TRF-2, Clone: 3H6B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP226298

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP226298

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 0.5% protein stabilizer
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7014
Conjugate Unlabeled
Cross Reactivity Human
Applications Immunohistochemistry
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Antigen TRF-2
Format Precipitation
Host Species Mouse
Immunogen Partial recombinant human TRF2 (between residues 300-500) expressed in &italic_begin;E. coli&italic_end; [Uniprot Q15554]
Quantity 0.1mL
Clone 3H6B4
Concentration 1.0mg/mL
Gene Accession No. Q15554