Please login to your online account to display your discounted pricing

anti-TRF-2, Clone: 3H6B4, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 3H6B4
Host Species Mouse
Isotype IgG1
Immunogen Partial recombinant human TRF2 (between residues 300-500) expressed in &italic_begin;E. coli&italic_end; [Uniprot Q15554]
Antigen TRF-2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP226298

 
novus biologicals canada
NBP226298
Unlabeled 0.1mL Each for $309.05

NBP226298SS

 
novus biologicals canada
NBP226298SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 3H6B4
Host Species Mouse
Isotype IgG1
Immunogen Partial recombinant human TRF2 (between residues 300-500) expressed in &italic_begin;E. coli&italic_end; [Uniprot Q15554]
Antigen TRF-2
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Western Blotting
Gene Accession No. Q15554
Gene ID (Entrez) 7014
Formulation PBS, 0.5% protein stabilizer
Concentration 1.0mg/mL