Please login to your online account to display your discounted pricing

anti-TRANCE/TNFSF11/RANK L, Clone: 12A668, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056593

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056593

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Immunogen A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen (NP_003692).
Quantity 0.1mg
Clone 12A668
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen TRANCE/TNFSF11/RANK L
Format Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1, κ
Gene Accession No. O14788
Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS (sterile filtered)
Regulatory Status RUO
Gene ID (Entrez) 8600
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal