Please login to your online account to display your discounted pricing

anti-TRANCE/TNFSF11/RANK L, Clone: 12A380, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 12A380
Host Species Mouse
Isotype IgG1, κ
Immunogen A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen.
Antigen TRANCE/TNFSF11/RANK L
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056748

 
novus biologicals canada
NB10056748
Unlabeled 0.1mg Each for $329.00

NB10056748T

 
novus biologicals canada
NB10056748SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 12A380
Host Species Mouse
Isotype IgG1, κ
Immunogen A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen.
Antigen TRANCE/TNFSF11/RANK L
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Gene ID (Entrez) 8600
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL