Please login to your online account to display your discounted pricing

anti-TRANCE/TNFSF11/RANK L, Clone: 12A380, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056748SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056748T

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Host Species Mouse
Antigen TRANCE/TNFSF11/RANK L
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Gene ID (Entrez) 8600
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Immunogen A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen.
Format Purified
Clone 12A380
Isotype IgG1, κ
Regulatory Status RUO
Quantity 0.025mg
Concentration 0.5mg/mL