Please login to your online account to display your discounted pricing

anti-TRANCE/TNFSF11/RANK L, Clone: 12A380, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056748

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056748

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Quantity 0.1mg
Antigen TRANCE/TNFSF11/RANK L
Format Purified
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Gene ID (Entrez) 8600
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Clone 12A380
Immunogen A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen.
Host Species Mouse
Monoclonal or Polyclonal Monoclonal