Please login to your online account to display your discounted pricing

anti-TRAIL R1/TNFRSF10A, Clone: 32A1380, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056747SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056747T

  • / Each
 

Description & Specifications

Specifications

Antigen TRAIL R1/TNFRSF10A
Applications Flow Cytometry
Applications Simple Western
Applications Western Blot
Clone 32A1380
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. O00220
Host Species Murine
Immunogen This monoclonal antibody was raised against a peptide corresponding to amino acid 2 to 21 of human DR4.
Isotype IgG1, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8797
Target Species Human