Please login to your online account to display your discounted pricing

anti-TRAF-5, Clone: 55A219, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 55A219
Host Species Mouse
Isotype IgG
Immunogen Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5.
Antigen TRAF-5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001107SS

 
novus biologicals canada
NB6001107SS
Unlabeled 0.025mg Each for $109.00

NB6001107

 
novus biologicals canada
NB6001107
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Clone 55A219
Host Species Mouse
Isotype IgG
Immunogen Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5.
Antigen TRAF-5
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Cross-reacts with Human. Not yet tested in other species.
Gene Accession No. O00463
Gene ID (Entrez) 7188
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL