Please login to your online account to display your discounted pricing

anti-TRAF-2, Clone: 33A1293, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056715

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056715

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS containing 0.05% BSA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2.
Monoclonal or Polyclonal Monoclonal
Clone 33A1293
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. Q12933
Primary or Secondary Primary
Gene ID (Entrez) 7186
Test Specificity In HeLa, an approx. 57 kDa band is observed.
Isotype IgG1, κ
Regulatory Status RUO
Host Species Mouse
Quantity 0.1mg
Antigen TRAF-2