Please login to your online account to display your discounted pricing

anti-TRAF-2, Clone: 33A1293, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 33A1293
Host Species Mouse
Isotype IgG1, κ
Immunogen Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2.
Antigen TRAF-2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056715

 
novus biologicals canada
NB10056715
Unlabeled 0.1mg Each for $329.00

NB10056715T

 
novus biologicals canada
NB10056715SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 33A1293
Host Species Mouse
Isotype IgG1, κ
Immunogen Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2.
Antigen TRAF-2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In HeLa, an approx. 57 kDa band is observed.
Gene Accession No. Q12933
Gene ID (Entrez) 7186
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL