Please login to your online account to display your discounted pricing

anti-TRAF-2, Clone: 33A1293, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056715SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056715T

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1, κ
Quantity 0.025mg
Gene ID (Entrez) 7186
Test Specificity In HeLa, an approx. 57 kDa band is observed.
Conjugate Unlabeled
Gene Accession No. Q12933
Primary or Secondary Primary
Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone 33A1293
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Format Purified
Antigen TRAF-2
Immunogen Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2.
Host Species Mouse
Regulatory Status RUO