Please login to your online account to display your discounted pricing

anti-TRA-1-81, Clone: TRA-1-81, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone TRA-1-81
Host Species Mouse
Isotype IgM
Immunogen Human embryonal carcinoma cell line 2102Ep.
Antigen TRA-1-81
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001833

 
novus biologicals canada
NB1001833
Unlabeled 0.1mL Each for $309.05

NB1001833SS

 
novus biologicals canada
NB1001833SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone TRA-1-81
Host Species Mouse
Isotype IgM
Immunogen Human embryonal carcinoma cell line 2102Ep.
Antigen TRA-1-81
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Human. This antibody recognizes the TRA-1-81 antigen that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). No immunoreactivity is seen with murine EC, EG or ES cells. This antibody recognizes antigens that are associated with a pericellular matrix proteoglycan.
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL