Please login to your online account to display your discounted pricing

anti-TRA-1-60 (R), Clone: TRA-1-60(R), Novus Biologicals™


Manufacturer: novus biologicals canada  NB100730SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100730SS

  • / Each
 

Description & Specifications

Specifications

Antigen TRA-1-60 (R)
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blot
Clone TRA-1-60(R)
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Tris-glycine, 150mM NaCl
Host Species Murine
Immunogen Human embryonal carcinoma cell line 2102Ep.
Isotype IgM
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Specific for the neuraminidase resistant form of the TRA-1-60 antigen that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). No immunoreactivity is seen with murine EC, EG or ES cells. TRA-1-60 reacts with a sialidase-sensitive epitope. Human. No immunoreactivity is seen with murine EC, EG or ES cells.
Target Species Human