missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse TNF-alpha, Polyclonal, R&D Systems

$467.98 - $481.01

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P06804
Gene ID (Entrez) 21926
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AB410NA R & D Systems
RDSAB410NA
1mg Each for $467.98
AF410NA R & D Systems
RDSAF410NA
100μg Each for $481.01
Specifications

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse TNF-α, Leu80-Leu235, Accession No. P06804

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P06804
21926