Please login to your online account to display your discounted pricing

anti-TNF-alpha, Clone: 6N1E7, Novus Biologicals™

$182.00 - $334.12

Specifications

Clone 6N1E7
Host Species Mouse
Immunogen Full-length recombinant human TNF alpha was used as the immunogen for this antibody.
Antigen TNF-alpha
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227224

 
novus biologicals canada
NBP227224
Unlabeled 0.1mg Each for $334.12

NBP227224SS

 
novus biologicals canada
NBP227224SS
Unlabeled 0.025mg Each for $182.00
Description & Specifications

Specifications

Clone 6N1E7
Host Species Mouse
Immunogen Full-length recombinant human TNF alpha was used as the immunogen for this antibody.
Antigen TNF-alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications Flow Cytometry
Applications Neutralization
Applications Western Blotting
Gene Accession No. P01375
Gene ID (Entrez) 7124
Formulation 0.1mg antibody in 0.2mL PBS; product is sterile.
Concentration 0.5mg/mL