Please login to your online account to display your discounted pricing

anti-TLR9, Clone: 26C593.2, Novus Biologicals™

$407.00 - $1,652.95

Specifications

Clone 26C593.2
Host Species Mouse
Isotype κ
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037).
Antigen TLR9
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224863

 
novus biologicals canada
NBP224863
Unlabeled 0.1mg Each for $407.00

NBP2248631

 
novus biologicals canada
NBP2248631MG
Unlabeled 1mg Each for $1,652.95
Description & Specifications

Specifications

Clone 26C593.2
Host Species Mouse
Isotype κ
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037).
Antigen TLR9
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This antibody is reactive to Dog, Horse, Human, Mouse, Rat and Rhesus Monkey.
Gene Accession No. Q9NR96
Gene ID (Entrez) 54106
Formulation Antibody in PBS. Filter sterilized.
Concentration 0.5mg/mL