Please login to your online account to display your discounted pricing

anti-TLR9, Clone: 26C593.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2248631MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2248631

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9NR96
Gene ID (Entrez) 54106
Conjugate Unlabeled
Primary or Secondary Primary
Concentration 0.5mg/mL
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 268-300 of human TLR9 isoform A (Genbank accession no. AAF78037).
Antigen TLR9
Isotype κ
Host Species Mouse
Quantity 1mg
Clone 26C593.2
Monoclonal or Polyclonal Monoclonal
Formulation Antibody in PBS. Filter sterilized.
Test Specificity This antibody is reactive to Dog, Horse, Human, Mouse, Rat and Rhesus Monkey.
Format Purified
Regulatory Status RUO