Please login to your online account to display your discounted pricing

anti-TLR5, Clone: 19D759.2, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 19D759.2
Host Species Mouse
Isotype κ
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 between amino acids 700-800. The antibody also reacts with mouse TLR5.
Antigen TLR5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224787

 
novus biologicals canada
NBP224787
Unlabeled 0.1mg Each for $329.00

NBP224787SS

 
novus biologicals canada
NBP224787SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 19D759.2
Host Species Mouse
Isotype κ
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 between amino acids 700-800. The antibody also reacts with mouse TLR5.
Antigen TLR5
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This antibody is reactive to Dog, Human and Mouse.
Gene Accession No. O60602
Gene ID (Entrez) 7100
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL