Please login to your online account to display your discounted pricing

anti-TLR4, Clone: HTA125, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056723

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056723

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications In vitro Assay
Applications Neutralization
Quantity 0.1mg
Antigen TLR4
Host Species Mouse
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Cross Reactivity Canine
Cross Reactivity Human
Gene ID (Entrez) 7099
Test Specificity The has been shown to block the activation of monocytes with LPS (Paik, et al, 2003).
Isotype IgG2a
Format Purified
Gene Accession No. O00206
Immunogen This antibody was developed by immunizing mice with Ba/F3 cell line expressing human TLR4 cell surface antigen.
Regulatory Status RUO
Clone HTA125