Please login to your online account to display your discounted pricing

anti-TLR4, Clone: 76B357.1, Novus Biologicals™

$142.00 - $384.00

Specifications

Clone 76B357.1
Host Species Murine
Isotype IgG2b, κ
Immunogen This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Antigen TLR4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056566T

 
novus biologicals canada
NB10056566SS
Unlabeled 0.025mg Each for $142.00

NB10056566

 
novus biologicals canada
NB10056566
Unlabeled 0.1mg Each for $384.00
Description & Specifications

Specifications

Clone 76B357.1
Host Species Murine
Isotype IgG2b, κ
Immunogen This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Antigen TLR4
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow (Cell Surface)
Applications Flow Cytometry
Applications Flow Cytometry (Cell Surface)
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used has 100% identity in human, chimp, baboon, chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. Human, Mouse, Rat. Ground squirrel reactivity reported by a customer review. Predicted cross-reactivity based on sequence identity: Chimp (100%), Baboon (100%), Chinese Hamster (100%), Bovine (93%), Pig (93%). Mammal reactivity reported in scientific literature (PMID: 25130694).
Gene Accession No. O00206
Gene ID (Entrez) 7099
Formulation PBS containing 0.05% BSA.
Concentration 0.5mg/mL