Please login to your online account to display your discounted pricing

anti-TLR4, Clone: 76B357.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056566SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056566T

  • / Each
 

Description & Specifications

Specifications

Antigen TLR4
Applications Flow Cytometry
Applications Flow Cytometry (Cell Surface)
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 76B357.1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Squirrel
Format Purified
Formulation PBS containing 0.05% BSA.
Gene Accession No. O00206
Host Species Murine
Immunogen This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Isotype IgG2b, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7099
Test Specificity The amino acid sequence used has 100% identity in human, chimp, baboon, chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. Human, Mouse, Rat. Ground squirrel reactivity reported by a customer review. Predicted cross-reactivity based on sequence identity: Chimp (100%), Baboon (100%), Chinese Hamster (100%), Bovine (93%), Pig (93%). Mammal reactivity reported in scientific literature (PMID: 25130694).
Target Species Human
Target Species Mammalia
Target Species Murine
Target Species Rat
Target Species Squirrel