Please login to your online account to display your discounted pricing

anti-TLR4, Clone: 76B357.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056566

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056566

  • / Each
 

Description & Specifications

Specifications

Applications Flow (Cell Surface)
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Squirrel
Clone 76B357.1
Gene Accession No. O00206
Primary or Secondary Primary
Formulation PBS containing 0.05% BSA.
Concentration 0.5mg/mL
Gene ID (Entrez) 7099
Conjugate Unlabeled
Test Specificity The amino acid sequence used has 100% identity in human, chimp, baboon, chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. Human, Mouse, Rat. Ground squirrel reactivity reported by a customer review. Predicted cross-reactivity based on sequence identity: Chimp (100%), Baboon (100%), Chinese Hamster (100%), Bovine (93%), Pig (93%). Mammal reactivity reported in scientific literature (PMID: 25130694).
Immunogen This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg
Antigen TLR4
Isotype IgG2b, κ