Please login to your online account to display your discounted pricing

anti-TLR3, Biotin, Clone: 619F7.06, Novus Biologicals™

$406.00 - $536.00

Specifications

Clone 619F7.06
Host Species Murine
Isotype IgM
Immunogen Human recombinant TLR3 in eukaryotic cells (pUNO-hTLR3)
Antigen TLR3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0470B

 
novus biologicals canada
DDX0470B
Biotin 0.1mg Each for $536.00

DDX0470B005

 
novus biologicals canada
DDX0470B005MG
Biotin 0.05mg Each for $406.00
Description & Specifications

Specifications

Clone 619F7.06
Host Species Murine
Isotype IgM
Immunogen Human recombinant TLR3 in eukaryotic cells (pUNO-hTLR3)
Antigen TLR3
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Gene Accession No. NP_003256
Gene ID (Entrez) 7098
Formulation Antibody in Tris-NaCl pH 8