Please login to your online account to display your discounted pricing

anti-TLR3, Biotin, Clone: 619F7.06, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0470B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0470B005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7098
Regulatory Status RUO
Clone 619F7.06
Cross Reactivity Human
Isotype IgM
Format Ion Exchange Chromatography
Immunogen Human recombinant TLR3 in eukaryotic cells (pUNO-hTLR3)
Monoclonal or Polyclonal Monoclonal
Applications Flow Cytometry
Applications Western Blotting
Gene Accession No. NP_003256
Primary or Secondary Primary
Host Species Mouse
Antigen TLR3
Quantity 0.05mg
Formulation Antibody in Tris-NaCl pH 8
Conjugate Biotin