Please login to your online account to display your discounted pricing

anti-TLR2, Clone: TL2.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056722

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056722

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Neutralization
Antigen TLR2
Quantity 0.1mg
Cross Reactivity Canine
Cross Reactivity Human
Formulation PBS containing, 0.05% BSA
Gene Accession No. O60603
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Format Purified
Concentration 0.5mg/mL
Host Species Mouse
Immunogen 2000). The hybridoma supernatants were selected by flow cytometry.
Isotype IgG2a
Clone TL2.1
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 7097