Please login to your online account to display your discounted pricing

anti-TLR2, Clone: 67N8F8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225245

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225245

  • / Each
 

Description & Specifications

Specifications

Antigen TLR2
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 67N8F8
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg of antibody in 0.2mL PBS containing, 0.05% BSA
Gene Accession No. NP_003255.2
Host Species Murine
Immunogen Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7097
Test Specificity This product reacts with Human.
Target Species Human