Please login to your online account to display your discounted pricing

anti-TLR2, Clone: 1030A5.138, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056728

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056728

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Formulation PBS containing 0.05% BSA
Antigen TLR2
Format Ascites
Gene ID (Entrez) 7097
Regulatory Status RUO
Primary or Secondary Primary
Conjugate Unlabeled
Quantity 0.2mL
Clone 1030A5.138
Cross Reactivity Human
Host Species Mouse
Immunogen This antibody was developed against extracellular domain of human TLR2.
Gene Accession No. O60603
Monoclonal or Polyclonal Monoclonal
Test Specificity In Ramos, an 86 kDa band is observed.