Please login to your online account to display your discounted pricing

anti-TLR2, Clone: 1030A5.138, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 1030A5.138
Host Species Mouse
Isotype IgG1
Immunogen This antibody was developed against extracellular domain of human TLR2.
Antigen TLR2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056728T

 
novus biologicals canada
NB10056728SS
Unlabeled 0.05mL Each for $139.00

NB10056728

 
novus biologicals canada
NB10056728
Unlabeled 0.2mL Each for $329.00
Description & Specifications

Specifications

Clone 1030A5.138
Host Species Mouse
Isotype IgG1
Immunogen This antibody was developed against extracellular domain of human TLR2.
Antigen TLR2
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In Ramos, an 86 kDa band is observed.
Gene Accession No. O60603
Gene ID (Entrez) 7097
Formulation PBS containing 0.05% BSA