Please login to your online account to display your discounted pricing

anti-TLR2, Clone: 1030A5.138, Novus Biologicals™

Manufacturer: novus biologicals canada  NB100-56728SS

 View all options for this product


For Research Use Only

Catalog No. NB10056728T


Description & Specifications


Antigen TLR2
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone 1030A5.138
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Formulation PBS containing 0.05% BSA
Gene Accession No. O60603
Host Species Mouse
Immunogen This antibody was developed against extracellular domain of human TLR2.
Isotype IgG1
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7097
Test Specificity In Ramos, an 86 kDa band is observed.