Please login to your online account to display your discounted pricing

anti-TLR12, Clone: 15F1215, Novus Biologicals™

$139.00 - $384.00

Specifications

Clone 15F1215
Host Species Murine
Isotype IgM
Immunogen This antibody was developed against amino acids 791-821 of mouse TLR12; GenBank NP_991392.1
Antigen TLR12
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224870SS

 
novus biologicals canada
NBP224870SS
Unlabeled 0.05mL Each for $139.00

NBP224870

 
novus biologicals canada
NBP224870
Unlabeled 0.2mL Each for $384.00
Description & Specifications

Specifications

Clone 15F1215
Host Species Murine
Isotype IgM
Immunogen This antibody was developed against amino acids 791-821 of mouse TLR12; GenBank NP_991392.1
Antigen TLR12
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This antibody is reactive to Mouse.
Gene ID (Entrez) 100131451
Formulation Ascites in 200μL PBS containing 0.2% gelatin