Please login to your online account to display your discounted pricing

anti-TLR10, Clone: 3C10C5, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 3C10C5
Host Species Murine
Isotype κ
Immunogen A partial recombinant protein containing amino acids 20-474 (extracellular domain) of human TLR10.
Antigen TLR10
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP170343

 
novus biologicals canada
NBP170343
Unlabeled 0.1mg Each for $307.00

NBP170343SS

 
novus biologicals canada
NBP170343SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 3C10C5
Host Species Murine
Isotype κ
Immunogen A partial recombinant protein containing amino acids 20-474 (extracellular domain) of human TLR10.
Antigen TLR10
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Gene Accession No. Q9BXR5
Gene ID (Entrez) 81793
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL