Please login to your online account to display your discounted pricing

anti-TLR10, Clone: 28M7A3, Novus Biologicals™

$192.00 - $406.00

Specifications

Clone 28M7A3
Host Species Mouse
Isotype κ
Immunogen A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody.
Antigen TLR10
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227273SS

 
novus biologicals canada
NBP227273SS
Unlabeled 0.025mg Each for $192.00

NBP227273

 
novus biologicals canada
NBP227273
Unlabeled 0.1mg Each for $406.00
Description & Specifications

Specifications

Clone 28M7A3
Host Species Mouse
Isotype κ
Immunogen A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody.
Antigen TLR10
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Gene Accession No. Q9BXR5
Gene ID (Entrez) 81793
Formulation 0.1mg of antibody in 0.2mL PBS, sterile filtered.
Concentration 0.5mg/mL