Please login to your online account to display your discounted pricing

anti-Tirap, Clone: 20D1055.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056730

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056730

  • / Each
 

Description & Specifications

Specifications

Antigen Tirap
Applications Western Blot
Clone 20D1055.1
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Formulation ascites fluid containing 0.05% BSA
Gene Accession No. P58753
Host Species Murine
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human TIRAP.
Isotype IgM, κ
Quantity 0.2mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 114609
Test Specificity In HUVEC, a 32 kDa band is observed.
Target Species Human