Please login to your online account to display your discounted pricing

anti-Tirap, Clone: 20D1055.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056730SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056730T

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Quantity 0.05mL
Formulation ascites fluid containing 0.05% BSA
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human TIRAP.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity In HUVEC, a 32 kDa band is observed.
Conjugate Unlabeled
Gene ID (Entrez) 114609
Isotype IgM, κ
Antigen Tirap
Format Ascites
Gene Accession No. P58753
Host Species Mouse
Regulatory Status RUO
Clone 20D1055.1
Cross Reactivity Human