Please login to your online account to display your discounted pricing

anti-TIN2, Clone: 59B388.1, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 59B388.1
Host Species Murine
Isotype IgG1
Immunogen A synthetic peptide corresponding to amino acids 44-58 of human Tin2 was used as immunogen (NP_036593.2).
Antigen TIN2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001522

 
novus biologicals canada
NB6001522
Unlabeled 0.1mg Each for $329.00

NB6001522SS

 
novus biologicals canada
NB6001522SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 59B388.1
Host Species Murine
Isotype IgG1
Immunogen A synthetic peptide corresponding to amino acids 44-58 of human Tin2 was used as immunogen (NP_036593.2).
Antigen TIN2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Gel Supershift
Applications Western Blot
Gene Accession No. Q9BSI4
Gene ID (Entrez) 26277
Formulation PBS with 0.05% BSA
Concentration 0.5mg/mL