Please login to your online account to display your discounted pricing

anti-TIN2, Clone: 59B388.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001522SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001522SS

  • / Each
 

Description & Specifications

Specifications

Antigen TIN2
Applications Gel Supershift
Applications Western Blot
Clone 59B388.1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS with 0.05% BSA
Gene Accession No. Q9BSI4
Host Species Murine
Immunogen A synthetic peptide corresponding to amino acids 44-58 of human Tin2 was used as immunogen (NP_036593.2).
Isotype IgG1
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 26277
Target Species Human
Target Species Murine