Please login to your online account to display your discounted pricing

anti-TIN2, Clone: 59B388.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001522

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001522

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Gel Supershift
Applications Western Blotting
Isotype IgG1
Quantity 0.1mg
Clone 59B388.1
Gene Accession No. Q9BSI4
Primary or Secondary Primary
Gene ID (Entrez) 26277
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Immunogen A synthetic peptide corresponding to amino acids 44-58 of human Tin2 was used as immunogen (NP_036593.2).
Regulatory Status RUO
Antigen TIN2