Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: DF1513, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232945

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232945

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Gene Accession No. P02786
Conjugate Unlabeled
Antigen TfR (Transferrin R)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen KG1 acute myeloid leukemia cell line
Clone DF1513
Isotype κ
Quantity 0.5mL
Primary or Secondary Primary
Gene ID (Entrez) 7037
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Cross Reactivity Human
Regulatory Status RUO
Format Purified
Host Species Mouse