Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: DF1513, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone DF1513
Host Species Mouse
Isotype κ
Immunogen KG1 acute myeloid leukemia cell line
Antigen TfR (Transferrin R)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232945

 
novus biologicals canada
NBP232945
Unlabeled 0.5mL Each for $321.00

NBP2329451M

 
novus biologicals canada
NBP2329451ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone DF1513
Host Species Mouse
Isotype κ
Immunogen KG1 acute myeloid leukemia cell line
Antigen TfR (Transferrin R)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P02786
Gene ID (Entrez) 7037
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL