Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: DF1513, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329451ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329451M

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype κ
Regulatory Status RUO
Antigen TfR (Transferrin R)
Format Purified
Cross Reactivity Human
Clone DF1513
Conjugate Unlabeled
Gene ID (Entrez) 7037
Formulation PBS with 0.05% BSA.
Gene Accession No. P02786
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Immunogen KG1 acute myeloid leukemia cell line
Quantity 1mL