Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: 66IG10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232946

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232946

  • / Each
 

Description & Specifications

Specifications

Antigen TfR (Transferrin R)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Clone 66IG10
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P02786
Host Species Mouse
Immunogen Human CD71
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7037