Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: 66IG10, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 66IG10
Host Species Mouse
Isotype κ
Immunogen Human CD71
Antigen TfR (Transferrin R)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329461M

 
novus biologicals canada
NBP2329461ML
Unlabeled 1mL Each for $429.00

NBP232946

 
novus biologicals canada
NBP232946
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone 66IG10
Host Species Mouse
Isotype κ
Immunogen Human CD71
Antigen TfR (Transferrin R)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Gene Accession No. P02786
Gene ID (Entrez) 7037
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL