Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: 66IG10, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2329461ML

 View all options for this product


For Research Use Only

Catalog No. NBP2329461M


Description & Specifications


Antigen TfR (Transferrin R)
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Clone 66IG10
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P02786
Host Species Mouse
Immunogen Human CD71
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7037