Please login to your online account to display your discounted pricing

anti-TERF21P, Clone: 78B356.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056526SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056526T

  • / Each
 

Description & Specifications

Specifications

Antigen TERF21P
Applications Western Blot
Clone 78B356.1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9NYB0
Host Species Murine
Immunogen A synthetic peptide corresponding amino acids 212-235 of human Rap1 was used to immunize mice.
Isotype IgG1
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 54386
Target Species Human