Please login to your online account to display your discounted pricing

anti-Tau, Clone: 2E9, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 2.00E+09
Host Species Mouse
Isotype IgG1
Immunogen Recombinant construct expressed in and purified from &italic_begin;E. coli&italic_end; which corresponded to the shortest version of the various Tau proteins
Antigen Tau
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225162

 
novus biologicals canada
NBP225162
Unlabeled 0.1mL Each for $309.05

NBP225162SS

 
novus biologicals canada
NBP225162SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 2.00E+09
Host Species Mouse
Isotype IgG1
Immunogen Recombinant construct expressed in and purified from &italic_begin;E. coli&italic_end; which corresponded to the shortest version of the various Tau proteins
Antigen Tau
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity This antibody is expected to bind to all Tau isoforms. Human, Mouse and Rat. Other species have not been tested.
Gene Accession No. P10636
Gene ID (Entrez) 4137
Formulation PBS
Concentration 1mg/mL