Please login to your online account to display your discounted pricing

anti-TAG-72, Clone: CA72/733, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329031ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329031M

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Quantity 1mL
Host Species Mouse
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Concentration 0.2mg/mL
Format Purified
Clone CA72/733
Regulatory Status RUO
Antigen TAG-72
Immunogen Recombinant human TAG-72 protein