Please login to your online account to display your discounted pricing

anti-TAG-72, Clone: CA72/733, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone CA72/733
Host Species Mouse
Isotype κ
Immunogen Recombinant human TAG-72 protein
Antigen TAG-72
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329031M

 
novus biologicals canada
NBP2329031ML
Unlabeled 1mL Each for $429.00

NBP232903

 
novus biologicals canada
NBP232903
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone CA72/733
Host Species Mouse
Isotype κ
Immunogen Recombinant human TAG-72 protein
Antigen TAG-72
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL