Please login to your online account to display your discounted pricing

anti-TAG-72, Clone: CA72/733, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232903

 View all options for this product


For Research Use Only

Catalog No. NBP232903


Description & Specifications


Antigen TAG-72
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone CA72/733
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Recombinant human TAG-72 protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal