Please login to your online account to display your discounted pricing

anti-Synuclein-alpha (Nitrate Tyr125, Nitrate Tyr133), Clone: 24.8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP126380

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP126380

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Isotype IgG1, κ
Antigen Nitrate Tyr133)
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL
Gene Accession No. P37840
Regulatory Status RUO
Host Species Mouse
Quantity 0.1mL
Immunogen Nitrated human alpha-Synuclein [UniProt No. P37840]
Clone 24.8
Gene ID (Entrez) 6622
Monoclonal or Polyclonal Monoclonal