Please login to your online account to display your discounted pricing

anti-Synuclein-alpha (Nitrate Tyr125, Nitrate Tyr133), Clone: 24.8, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone 24.8
Host Species Mouse
Isotype IgG1, κ
Immunogen Nitrated human alpha-Synuclein [UniProt No. P37840]
Antigen Nitrate Tyr133)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP126380SS

 
novus biologicals canada
NBP126380SS
Unlabeled 0.025mL Each for $109.00

NBP126380

 
novus biologicals canada
NBP126380
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone 24.8
Host Species Mouse
Isotype IgG1, κ
Immunogen Nitrated human alpha-Synuclein [UniProt No. P37840]
Antigen Nitrate Tyr133)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P37840
Gene ID (Entrez) 6622
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL